siRNA Details
| Gene Symbol: Ldlrap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145554.2 | NP_663529.2 | 9 | 934 | ||
| GenBank mRNA | AK029008.1 | - | 9 | 899 | ||
| AK148340.1 | - | 10 | 896 | |||
| AK159800.1 | - | 9 | 878 | |||
| AK167290.1 | - | 9 | 883 | |||
| AK169675.1 | - | 4 | 367 | |||
| BC021467.1 | - | 9 | 883 | 5067154 | ||
| BC066808.1 | - | 9 | 877 | 30055031 | ||
| BC067411.1 | - | 9 | 877 | 30092248 | ||

Ldlrap1
Genomic Map