siRNA Details
| Gene Symbol: Rcc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001197082.1 | NP_001184011.1 | 7 | 772 | ||
| NM_133878.3 | NP_598639.1 | 6 | 733 | |||
| GenBank mRNA | AK076089.1 | - | 6 | 734 | ||
| AK150153.1 | - | 6 | 685 | |||
| AK150610.1 | - | 6 | 683 | |||
| AK153193.1 | - | 6 | 683 | |||
| AK155225.1 | - | 4 | 315 | |||
| BC019807.1 | - | 6 | 645 | 4456795 | ||
| BC057645.1 | - | 7 | 727 | 3591859 | ||

Rcc1
Genomic Map