siRNA Details
| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153565.2 | NP_705793.1 | 4 | 965 | ||
| GenBank mRNA | AK043077.1 | - | 4 | 815 | ||
| AK149520.1 | - | 4 | 744 | |||
| AY273821.1 | - | 4 | 589 | |||
| BC038085.1 | - | 4 | 980 | 4502199 | ||

Pcsk9
Genomic Map