siRNA Details
| Gene Symbol: Pcsk9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153565.2 | NP_705793.1 | 12 | 2318 | ||
| GenBank mRNA | AK043077.1 | - | 12 | 2167 | ||
| AK149520.1 | - | 12 | 2097 | |||
| AY273821.1 | - | 12 | 1942 | |||
| BC038085.1 | - | 12 | 2333 | 4502199 | ||

Pcsk9
Genomic Map