siRNA Details
| Gene Symbol: Zmym6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001285885.1 | NP_001272814.1 | 14 | 3362 | ||
| NM_001285887.1 | NP_001272816.1 | 14 | 2279 | |||
| NM_001285888.1 | NP_001272817.1 | 15 | 2562 | |||
| NM_177462.5 | NP_803413.1 | 15 | 2555 | |||
| GenBank mRNA | AK035136.1 | - | 15 | 2557 | ||
| AK081836.1 | - | 14 | 2269 | |||
| BC034119.1 | - | 13 | 2207 | |||
| BC040744.1 | - | 13 | 2207 | |||
| BC094598.1 | - | 14 | 3335 | |||
| BC140958.1 | - | 15 | 2430 | 9055570 | ||
| BC144807.1 | - | 15 | 2437 | |||

Zmym6
Genomic Map