siRNA Details
| Gene Symbol: Rusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037709.2 | NP_001032798.1 | 9 | 3653 | ||
| NM_199057.3 | NP_951012.2 | 9 | 3614 | |||
| GenBank mRNA | AK053542.1 | - | 5 | 2523 | ||
| AK122263.1 | - | 9 | 3533 | |||
| AK132170.1 | - | 9 | 3543 | |||
| AK166260.1 | - | 9 | 3554 | |||
| BC024790.1 | - | 7 | 844 | |||
| BC141186.1 | - | 8 | 3384 | 9055798 | ||

Rusc2
Genomic Map