siRNA Details
| Gene Symbol: Rusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037709.2 | NP_001032798.1 | 4, 5 | 3139 | ||
| NM_199057.3 | NP_951012.2 | 4, 5 | 3100 | |||
| GenBank mRNA | AK122263.1 | - | 4, 5 | 3019 | ||
| AK132170.1 | - | 4, 5 | 3029 | |||
| BC024790.1 | - | 2, 3 | 330 | |||

Rusc2
Genomic Map