siRNA Details
| Gene Symbol: Rusc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037709.2 | NP_001032798.1 | 3 | 2504 | ||
| NM_199057.3 | NP_951012.2 | 3 | 2465 | |||
| GenBank mRNA | AK053542.1 | - | 2 | 483 | ||
| AK122263.1 | - | 3 | 2384 | |||
| AK132170.1 | - | 3 | 2394 | |||
| AK166260.1 | - | 3 | 2405 | |||
| BC141186.1 | - | 2 | 2235 | 9055798 | ||

Rusc2
Genomic Map