siRNA Details
| Gene Symbol: Osbpl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134791.2 | NP_001128263.1 | 8, 9 | 716 | ||
| NM_133885.4 | NP_598646.4 | 11, 12 | 949 | |||
| NM_173350.3 | NP_775485.2 | 9, 10 | 755 | |||
| GenBank mRNA | AK035658.1 | - | 9, 10 | 704 | ||
| AK131984.1 | - | 11, 12 | 920 | |||
| AK172010.1 | - | 9, 10 | 742 | |||
| AK215553.1 | - | 2, 3 | 256 | |||
| BC021507.1 | - | 7, 8 | 577 | 5347121 | ||
| BC023759.1 | - | 11, 12 | 904 | |||
| BC026927.1 | - | 9, 10 | 638 | 5134780 | ||
| BC089163.1 | - | 8, 9 | 657 | 5367603 | ||

Osbpl9
Genomic Map