siRNA Details
| Gene Symbol: AU040320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035525.1 | NP_001030602.1 | 13 | 2165 | ||
| NM_001035526.1 | NP_001030603.1 | 13 | 2283 | |||
| NM_133886.2 | NP_598647.1 | 13 | 2283 | |||
| GenBank mRNA | AK031342.1 | - | 13 | 2227 | ||
| AK043006.1 | - | 13 | 2233 | |||
| AK049570.1 | - | 13 | 2166 | |||
| AK084668.1 | - | 13 | 2128 | |||
| AK147569.1 | - | 4814 | ||||
| AK161493.1 | - | 3 | 346 | |||
| AK170261.1 | - | 13 | 2240 | |||
| AK198153.1 | - | 3 | 281 | |||
| AK211858.1 | - | 3 | 366 | |||
| BC022154.1 | - | 4 | 695 | |||
| BC028869.1 | - | 13 | 2253 | 4235813 | ||

AU040320
Genomic Map