siRNA Details
| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159633.1 | NP_001153105.1 | 10 | 1503 | ||
| NM_133891.3 | NP_598652.3 | 10 | 1503 | |||
| GenBank mRNA | AK077951.1 | - | 10 | 1506 | ||
| AY249865.1 | - | 10 | 1259 | |||
| AY249866.1 | - | 10 | 1232 | |||
| BC025941.1 | - | 1 | 65 | |||
| BC113167.1 | - | 10 | 1259 | 40059854 | ||
| BC113169.1 | - | 10 | 1259 | 40059860 | ||

Slc44a1
Genomic Map