siRNA Details
| Gene Symbol: Slc44a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133891.3 | NP_598652.3 | 14 | 2049 | ||
| GenBank mRNA | AK014708.1 | - | 1 | 302 | ||
| AY249865.1 | - | 14 | 1805 | |||
| AY249866.1 | - | 14 | 1778 | |||
| BC010258.1 | - | 2 | 243 | |||
| BC025941.1 | - | 5 | 611 | |||
| BC113167.1 | - | 14 | 1805 | 40059854 | ||
| BC113169.1 | - | 14 | 1805 | 40059860 | ||

Slc44a1
Genomic Map