siRNA Details
| Gene Symbol: Zfp518b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081144.2 | NP_001074613.2 | 4 | 2383 | ||
| NM_001177902.1 | NP_001171373.1 | 3 | 2317 | |||
| GenBank mRNA | AK173245.1 | - | 1 | 1361 | ||
| BC138368.1 | - | 1 | 1742 | 8861388 | ||
| BC138369.1 | - | 1 | 1802 | 8861389 | ||

Zfp518b
Genomic Map