siRNA Details
| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081422.2 | NP_001074891.1 | 10 | 6253 | ||
| GenBank mRNA | AF013969.1 | - | 1 | 1712 | ||
| AK122495.1 | - | 1 | 1397 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bod1l| Gene Symbol: Bod1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081422.2 | NP_001074891.1 | 10 | 6253 | ||
| GenBank mRNA | AF013969.1 | - | 1 | 1712 | ||
| AK122495.1 | - | 1 | 1397 | |||