siRNA Details
| Gene Symbol: Acacb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133904.2 | NP_598665.2 | 42 | 5994 | ||
| GenBank mRNA | AK076301.1 | - | 1 | 84 | ||
| AK147463.1 | - | 31 | 4189 | |||
| AY451394.1 | - | 42 | 5994 | |||
| BC022940.1 | - | 8 | 1080 | |||

Acacb
Genomic Map