siRNA Details
Gene Symbol: Acacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133904.2 | NP_598665.2 | 1 | 583 | ||
GenBank mRNA | AF290178.2 | - | 1 | 523 | ||
AY451394.1 | - | 1 | 583 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Acacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133904.2 | NP_598665.2 | 1 | 583 | ||
GenBank mRNA | AF290178.2 | - | 1 | 523 | ||
AY451394.1 | - | 1 | 583 |