siRNA Details
| Gene Symbol: Ube3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133907.3 | NP_598668.1 | 19 | 2779 | ||
| GenBank mRNA | AK029973.1 | - | 19 | 2589 | ||
| AK046056.1 | - | 19 | 2780 | |||
| AK049125.1 | - | 2 | 170 | |||
| AK122187.1 | - | 19 | 2714 | |||
| AK185151.1 | - | 3 | 149 | |||
| AK203997.1 | - | 2 | 250 | |||
| BC021525.1 | - | 8 | 1024 | |||
| BC059245.1 | - | 19 | 2594 | |||
| BC120731.1 | - | 19 | 2526 | 40129654 | ||
| BC137626.1 | - | 19 | 2526 | 8860645 | ||

Ube3c
Genomic Map