siRNA Details
| Gene Symbol: Emilin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133918.2 | NP_598679.1 | 1 | 125 | ||
| GenBank mRNA | AK020015.1 | - | 1 | 172 | ||
| AK029337.1 | - | 1 | 96 | |||
| AK052368.1 | - | 1 | 206 | |||
| AK088995.1 | - | 1 | 188 | |||
| AK138020.1 | - | 1 | 147 | |||
| AK155486.1 | - | 1 | 427 | |||
| BC005481.1 | - | 1 | 125 | 3485057 | ||

Emilin1
Genomic Map