siRNA Details
| Gene Symbol: Hepacam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178899.5 | NP_849230.1 | 1, 2 | 93 | ||
| GenBank mRNA | AK033728.1 | - | 1, 2 | 95 | ||
| AK050501.1 | - | 1, 2 | 94 | |||
| BC096374.1 | - | 1, 2 | 80 | 4483627 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hepacam2| Gene Symbol: Hepacam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178899.5 | NP_849230.1 | 1, 2 | 93 | ||
| GenBank mRNA | AK033728.1 | - | 1, 2 | 95 | ||
| AK050501.1 | - | 1, 2 | 94 | |||
| BC096374.1 | - | 1, 2 | 80 | 4483627 | ||