siRNA Details
| Gene Symbol: Hepacam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178899.5 | NP_849230.1 | 2 | 337 | ||
| GenBank mRNA | AK033728.1 | - | 2 | 339 | ||
| AK050501.1 | - | 2 | 338 | |||
| BC096374.1 | - | 2 | 324 | 4483627 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hepacam2| Gene Symbol: Hepacam2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178899.5 | NP_849230.1 | 2 | 337 | ||
| GenBank mRNA | AK033728.1 | - | 2 | 339 | ||
| AK050501.1 | - | 2 | 338 | |||
| BC096374.1 | - | 2 | 324 | 4483627 | ||