siRNA Details
| Gene Symbol: Phrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081118.1 | NP_001074587.1 | 14 | 3377 | ||
| GenBank mRNA | AK129387.1 | - | 1 | 1803 | ||
| AK134140.1 | - | 1 | 944 | |||
| AK161749.1 | - | 1 | 478 | |||
| AK164438.1 | - | 1 | 250 | |||
| BC094566.1 | - | 3 | 1852 | |||
| BC138446.1 | - | 9 | 3023 | 8861466 | ||
| BC145719.1 | - | 9 | 3023 | 40131022 | ||

Phrf1
Genomic Map