siRNA Details
| Gene Symbol: Plekha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133942.2 | NP_598703.1 | 5 | 481 | ||
| GenBank mRNA | AK044224.1 | - | 7 | 633 | ||
| AK083340.1 | - | 7 | 655 | |||
| BC020017.1 | - | 5 | 481 | 3589001 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Plekha1| Gene Symbol: Plekha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133942.2 | NP_598703.1 | 5 | 481 | ||
| GenBank mRNA | AK044224.1 | - | 7 | 633 | ||
| AK083340.1 | - | 7 | 655 | |||
| BC020017.1 | - | 5 | 481 | 3589001 | ||