siRNA Details
| Gene Symbol: Ric8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053194.4 | NP_444424.1 | 2, 3 | 207 | ||
| GenBank mRNA | AF288813.1 | - | 2, 3 | 206 | ||
| AK031466.1 | - | 2, 3 | 154 | |||
| AK080192.1 | - | 2, 3 | 150 | |||
| AK155282.1 | - | 2, 3 | 182 | |||
| AK165536.1 | - | 2, 3 | 184 | |||
| AK167746.1 | - | 2, 3 | 210 | |||
| AK169367.1 | - | 2, 3 | 171 | |||
| AK212668.1 | - | 1, 2 | 55 | |||
| BC022917.1 | - | 2, 3 | 184 | 4460521 | ||

Ric8
Genomic Map