siRNA Details
| Gene Symbol: Ccp110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182995.1 | NP_892040.1 | 5 | 1682 | ||
| GenBank mRNA | AK141641.1 | - | 5 | 1677 | ||
| AK172946.1 | - | 1 | 896 | |||
| BC053103.1 | - | 5 | 1682 | 6406475 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccp110| Gene Symbol: Ccp110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182995.1 | NP_892040.1 | 5 | 1682 | ||
| GenBank mRNA | AK141641.1 | - | 5 | 1677 | ||
| AK172946.1 | - | 1 | 896 | |||
| BC053103.1 | - | 5 | 1682 | 6406475 | ||