siRNA Details
| Gene Symbol: Numa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133947.3 | NP_598708.3 | 14 | 3967 | ||
| GenBank mRNA | BC049791.1 | - | 14 | 3967 | 6505037 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Numa1| Gene Symbol: Numa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133947.3 | NP_598708.3 | 14 | 3967 | ||
| GenBank mRNA | BC049791.1 | - | 14 | 3967 | 6505037 | |