siRNA Details
| Gene Symbol: Ano1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242349.1 | NP_001229278.1 | 19 | 2168 | ||
| NM_178642.5 | NP_848757.4 | 20 | 2177 | |||
| GenBank mRNA | AK028991.1 | - | 2 | 79 | ||
| AK029329.1 | - | 6 | 544 | |||
| AK032793.1 | - | 15 | 1189 | |||
| AK052589.1 | - | 20 | 2129 | |||
| BC004697.1 | - | 12 | 1043 | |||
| BC006062.1 | - | 3 | 127 | |||
| BC062959.1 | - | 19 | 2054 | 30547439 | ||

Ano1
Genomic Map