siRNA Details
| Gene Symbol: Ints4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027256.2 | NP_081532.1 | 3 | 431 | ||
| GenBank mRNA | AK088635.1 | - | 3 | 375 | ||
| AK163383.1 | - | 3 | 385 | |||
| BC013710.1 | - | 3 | 377 | 3156446 | ||
| CA450656.1 | - | 3 | 301 | |||

Ints4
Genomic Map