siRNA Details
| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133962.3 | NP_598723.3 | 15, 16 | 2083 | ||
| GenBank mRNA | AK147923.1 | - | 15, 16 | 2056 | ||
| AK172963.1 | - | 9, 10 | 1186 | |||
| BC034512.1 | - | 4, 5 | 342 | |||
| BC060639.1 | - | 15, 16 | 2055 | 6849875 | ||

Arhgef18
Genomic Map