siRNA Details
| Gene Symbol: Arhgef18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133962.3 | NP_598723.3 | 12 | 1751 | ||
| GenBank mRNA | AK041423.1 | - | 12 | 1754 | ||
| AK147923.1 | - | 12 | 1723 | |||
| AK172963.1 | - | 6 | 854 | |||
| BC060639.1 | - | 12 | 1723 | 6849875 | ||

Arhgef18
Genomic Map