siRNA Details
| Gene Symbol: Fam192a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028221.4 | NP_082497.2 | 8 | 1033 | ||
| GenBank mRNA | AK033988.1 | - | 8 | 964 | ||
| AK050704.1 | - | 8 | 968 | |||
| AK151603.1 | - | 8 | 987 | |||
| BC016088.1 | - | 7 | 773 | 4503452 | ||
| BC023158.1 | - | 8 | 872 | 4946201 | ||
| BC025472.1 | - | 8 | 1009 | 5353148 | ||
| BC027060.1 | - | 8 | 995 | 5358022 | ||
| BC027818.1 | - | 7 | 801 | 5320967 | ||
| BC040818.1 | - | 986 | 4194531 | |||

Fam192a
Genomic Map