siRNA Details
| Gene Symbol: Tnks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083986.1 | - | 1250 | |||
| BC046509.1 | - | 837 | ||||
| BC094602.1 | - | 936 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tnks| Gene Symbol: Tnks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK083986.1 | - | 1250 | |||
| BC046509.1 | - | 837 | ||||
| BC094602.1 | - | 936 | ||||