siRNA Details
Gene Symbol: Cog4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133973.2 | NP_598734.1 | 15 | 1863 | ||
GenBank mRNA | AK156466.1 | - | 15 | 1855 | ||
AK172465.1 | - | 15 | 1854 | |||
AY223866.1 | - | 15 | 1890 | |||
BC023120.1 | - | 15 | 1840 | 4161858 |
