siRNA Details
| Gene Symbol: Cog4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133973.2 | NP_598734.1 | 6, 7 | 859 | ||
| GenBank mRNA | AK156466.1 | - | 6, 7 | 850 | ||
| AK172465.1 | - | 6, 7 | 850 | |||
| AK193863.1 | - | 1, 2 | 27 | |||
| AY223866.1 | - | 6, 7 | 886 | |||
| BC023120.1 | - | 6, 7 | 836 | 4161858 | ||

Cog4
Genomic Map