siRNA Details
| Gene Symbol: Ano10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271873.1 | NP_001258802.1 | 7 | 1154 | ||
| NM_133979.3 | NP_598740.1 | 7 | 1328 | |||
| GenBank mRNA | AK028391.1 | - | 2 | 273 | ||
| AK043011.1 | - | 7 | 1291 | |||
| AK046091.1 | - | 7 | 1280 | |||
| AK054163.1 | - | 3 | 618 | |||
| AK079622.1 | - | 7 | 1279 | |||
| AK080947.1 | - | 7 | 1280 | |||
| AK138414.1 | - | 7 | 1403 | |||
| BC026421.1 | - | 2 | 368 | |||
| BC099688.1 | - | 7 | 1077 | 6313494 | ||

Ano10
Genomic Map