siRNA Details
| Gene Symbol: Snx19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028874.2 | NP_083150.1 | 1 | 1599 | ||
| GenBank mRNA | AK122231.1 | - | 1 | 1820 | ||
| BC004635.1 | - | 1 | 71 | |||
| BC051164.1 | - | 1 | 1819 | |||
| BC063262.1 | - | 1 | 1599 | 6405897 | ||

Snx19
Genomic Map