siRNA Details
Gene Symbol: Slc6a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142809.1 | NP_001136281.1 | 2 | 1021 | ||
NM_001142810.1 | NP_001136282.1 | 2 | 1021 | |||
NM_133987.2 | NP_598748.1 | 2 | 1021 | |||
GenBank mRNA | AB077327.1 | - | 2 | 369 | ||
AF459435.1 | - | 2 | 1004 | |||
AK180138.1 | - | 1 | 12 | |||
AK214959.1 | - | 1 | 12 | |||
BC024444.1 | - | 1 | 34 | |||
BC024564.1 | - | 1 | 168 | |||
BC049801.1 | - | 2 | 196 | |||
BC141067.1 | - | 3 | 762 | 9055679 | ||
BC145319.1 | - | 3 | 426 | 9053874 |
