siRNA Details
| Gene Symbol: D330045A20Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175326.5 | NP_780535.1 | 11, 12 | 2108 | ||
| GenBank mRNA | AK052376.1 | - | 11, 12 | 2032 | ||
| AK155588.1 | - | 11, 12 | 2105 | |||
| AK171459.1 | - | 11, 12 | 2104 | |||
| BC112416.1 | - | 10, 11 | 1916 | |||
| BC113128.1 | - | 11, 12 | 2008 | 40061080 | ||

D330045A20Rik
Genomic Map