siRNA Details
| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043344.1 | - | 18 | 1995 | ||
| AK145200.1 | - | 19 | 2051 | |||
| AK161473.1 | - | 19 | 2134 | |||
| BC024848.1 | - | 2 | 647 | |||
| BC030328.1 | - | 19 | 2051 | 5376020 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cenpi| Gene Symbol: Cenpi | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK043344.1 | - | 18 | 1995 | ||
| AK145200.1 | - | 19 | 2051 | |||
| AK161473.1 | - | 19 | 2134 | |||
| BC024848.1 | - | 2 | 647 | |||
| BC030328.1 | - | 19 | 2051 | 5376020 | ||