siRNA Details
| Gene Symbol: Pwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133993.3 | NP_598754.2 | 10 | 1025 | ||
| GenBank mRNA | AK009972.1 | - | 10 | 992 | ||
| AK011891.1 | - | 10 | 973 | |||
| AK012649.1 | - | 4 | 268 | |||
| AK075805.1 | - | 10 | 967 | |||
| AK082158.1 | - | 10 | 967 | |||
| AK149699.1 | - | 10 | 970 | |||
| BC003199.1 | - | 10 | 994 | 3586496 | ||
| BC023137.1 | - | 10 | 975 | 3986908 | ||

Pwp1
Genomic Map