siRNA Details
| Gene Symbol: Pwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133993.3 | NP_598754.2 | 4 | 467 | ||
| GenBank mRNA | AK009972.1 | - | 4 | 434 | ||
| AK011891.1 | - | 4 | 415 | |||
| AK075805.1 | - | 4 | 409 | |||
| AK082158.1 | - | 4 | 409 | |||
| AK149699.1 | - | 4 | 411 | |||
| BC003199.1 | - | 4 | 436 | 3586496 | ||
| BC023137.1 | - | 4 | 417 | 3986908 | ||

Pwp1
Genomic Map