siRNA Details
Gene Symbol: Ncln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134009.3 | NP_598770.1 | 12 | 1634 | ||
GenBank mRNA | AK048954.1 | - | 12 | 1628 | ||
AK181997.1 | - | 1 | 37 | |||
AK209854.1 | - | 1 | 37 | |||
BC019501.1 | - | 12 | 1634 | 4193809 |
