siRNA Details
| Gene Symbol: Nup107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134010.2 | NP_598771.1 | 7 | 724 | ||
| GenBank mRNA | AK043454.1 | - | 7 | 719 | ||
| AK088069.1 | - | 7 | 725 | |||
| AK088303.1 | - | 7 | 701 | |||
| AK088625.1 | - | 7 | 694 | |||
| AK133938.1 | - | 7 | 684 | |||
| AK161665.1 | - | 7 | 712 | |||
| BC004655.1 | - | 3 | 199 | |||
| BC057591.1 | - | 7 | 691 | 6402677 | ||
| BC092261.1 | - | 7 | 701 | 2646396 | ||

Nup107
Genomic Map