siRNA Details
| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035226.1 | NP_001030303.1 | 18 | 2190 | ||
| NM_134014.3 | NP_598775.2 | 18 | 2428 | |||
| GenBank mRNA | AK052007.1 | - | 1 | 176 | ||
| AK136538.1 | - | 11 | 1591 | |||
| AK142055.1 | - | 1 | 933 | |||
| BC012276.1 | - | 5 | 652 | 3156478 | ||
| BC025628.1 | - | 4 | 612 | 5321933 | ||
| BC029714.1 | - | 16 | 1951 | |||
| BC034710.1 | - | 17 | 2185 | |||
| BC062912.1 | - | 18 | 2190 | 6821729 | ||
| BQ831940.1 | - | 1 | 92 | |||
| BU052960.1 | - | 4 | 533 | |||

Xpo1
Genomic Map