siRNA Details
| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035226.1 | NP_001030303.1 | 24 | 3100 | ||
| NM_134014.3 | NP_598775.2 | 24 | 3338 | |||
| GenBank mRNA | AK037196.1 | - | 266 | |||
| AK136538.1 | - | 17 | 2501 | |||
| BC012276.1 | - | 11 | 1562 | 3156478 | ||
| BC025628.1 | - | 10 | 1522 | 5321933 | ||
| BC029714.1 | - | 22 | 2861 | |||
| BC034710.1 | - | 23 | 3095 | |||
| BC062912.1 | - | 24 | 3100 | 6821729 | ||

Xpo1
Genomic Map