siRNA Details
| Gene Symbol: Xpo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035226.1 | NP_001030303.1 | 17, 18 | 2085 | ||
| NM_134014.3 | NP_598775.2 | 17, 18 | 2323 | |||
| GenBank mRNA | AK136538.1 | - | 10, 11 | 1486 | ||
| BC012276.1 | - | 4, 5 | 547 | 3156478 | ||
| BC025628.1 | - | 3, 4 | 507 | 5321933 | ||
| BC029714.1 | - | 15, 16 | 1846 | |||
| BC034710.1 | - | 16, 17 | 2080 | |||
| BC062912.1 | - | 17, 18 | 2085 | 6821729 | ||
| BU052960.1 | - | 3, 4 | 428 | |||

Xpo1
Genomic Map