siRNA Details
| Gene Symbol: Slc35e4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153142.3 | NP_694782.1 | 1 | 1668 | ||
| GenBank mRNA | AK039786.1 | - | 1 | 1669 | ||
| BC020181.1 | - | 1 | 702 | 4488513 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc35e4| Gene Symbol: Slc35e4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153142.3 | NP_694782.1 | 1 | 1668 | ||
| GenBank mRNA | AK039786.1 | - | 1 | 1669 | ||
| BC020181.1 | - | 1 | 702 | 4488513 | ||