siRNA Details
Gene Symbol: Slc35e4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153142.3 | NP_694782.1 | 2 | 2140 | ||
GenBank mRNA | AK039786.1 | - | 2 | 2141 | ||
BC020181.1 | - | 2 | 1174 | 4488513 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc35e4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153142.3 | NP_694782.1 | 2 | 2140 | ||
GenBank mRNA | AK039786.1 | - | 2 | 2141 | ||
BC020181.1 | - | 2 | 1174 | 4488513 |