siRNA Details
| Gene Symbol: Pex12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134025.3 | NP_598786.1 | 2, 3 | 380 | ||
| GenBank mRNA | AK035552.1 | - | 2, 3 | 381 | ||
| AK041175.1 | - | 2, 3 | 474 | |||
| AK153798.1 | - | 1, 2 | 587 | |||
| AK166413.1 | - | 2, 3 | 424 | |||
| BC021800.1 | - | 1, 2 | 204 | 3671142 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053921, XM_006247026, XM_008767934, XM_008773868, XM_006246967, and others.

Pex12
Genomic Map