siRNA Details
| Gene Symbol: Plin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113471.1 | NP_001106942.1 | 5 | 440 | ||
| NM_175640.2 | NP_783571.2 | 5 | 496 | |||
| GenBank mRNA | AK027962.1 | - | 2 | 156 | ||
| AK031445.1 | - | 4 | 421 | |||
| AY161165.1 | - | 4 | 391 | |||
| BC096685.1 | - | 5 | 441 | 1448913 | ||
| BY723040.1 | - | 5 | 555 | |||

Plin1
Genomic Map