siRNA Details
| Gene Symbol: Qsox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024945.1 | NP_001020116.1 | 10 | 1362 | ||
| NM_023268.2 | NP_075757.1 | 10 | 1362 | |||
| GenBank mRNA | AB044284.1 | - | 10 | 1270 | ||
| AK004880.1 | - | 10 | 1308 | |||
| AK083938.1 | - | 10 | 1313 | |||
| AK166839.1 | - | 10 | 1308 | |||
| AK169877.1 | - | 10 | 1315 | |||
| AK170449.1 | - | 10 | 1275 | |||
| BC034131.1 | - | 7 | 854 | |||
| BC076590.1 | - | 10 | 1316 | 30548238 | ||

Qsox1
Genomic Map